Anti-TH (phospho-S8) Rabbit Polyclonal Antibody
77701-477
New Product
- Antigen Symbol:TH (phospho-S8)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P07101
- Antigen Synonyms:Protein Pale|ple|EC 1.14.16.2|DYT5b|TH|OTTHUMP00000011225|TYH|TY3H_HUMAN|OTTHUMP00000011226|Tyrosine 3 monooxygenase|Tyrosine 3-hydroxylase|Tyrosine 3 hydroxylase|The|Tyrosine 3-monooxygenase|DYT14|Dystonia 14|Tyrosine hydroxylase
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human TH around the phosphorylation site of S8
- Purification:Antigen affinity purified
- Cat. No.:77701-477
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: TH (phospho-S8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat