Anti-BMP15 Rabbit Polyclonal Antibody
77701-462
New Product
- Antigen Symbol:BMP15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O95972
- Antigen Synonyms:BMP15_HUMAN|BMP-15|ODG2 |BMP15|GDF-9B|GDF 9B|Growth/differentiation factor 9B|GDF9B|POF4|Bone morphogenetic protein 15|BMP 15
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human BMP-15
- Purification:Antigen affinity purified
- Cat. No.:77701-462
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: BMP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human