Anti-CD300C Rabbit Polyclonal Antibody
Catalog # 77701-431
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CD300C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q08708
- Antigen Synonyms:CMRF35|CLM-6|CMRF35-A1|Immunoglobulin superfamily member 16|CMRF35-like molecule 6|IgSF16|CMRF-35A|CMRF-35|CD300c|CMRF35 antigen|CMRF35A|CMRF35 leukocyte immunoglobulin-like receptor|CMRF35A1|CD300c molecule|CLM6_HUMAN|CD300 antigen-like family member C|CD300c antigen|LIR|CMRF35A leukocyte immunoglobulin-like receptor
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CD300c
- Purification:Antigen affinity purified
- Cat. No.:77701-431
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: CD300C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human