Anti-ALPL Rabbit Polyclonal Antibody
Catalog # 77701-375
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ALPL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Size:100 µg
- Form:Liquid
- Gene ID:P05186
- Antigen Synonyms:FLJ93059|Alpl|Tissue nonspecific ALP|Alkaline phosphatase tissue nonspecific isozyme|Tissue non specific alkaline phosphatase|BAP|Alkaline phosphatase liver/bone/kidney isozyme|Glycerophosphatase|AKP2|Liver/bone/kidney type alkaline phosphatase|MGC167935|MGC161443|PHOA|Alkaline phosphatase liver/bone/kidney|Alkaline phosphatase, tissue-nonspecific isozyme|TNSALP|AP-TNAP|AP TNAP|Alkaline phosphomonoesterase|TNAP|FLJ40094|HOPS|APTNAP|PPBT_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human TNAP
- Purification:Antigen affinity purified
- Cat. No.:77701-375
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ALPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat