Anti-HLA-DQB1/HLA-DQB2 Rabbit Polyclonal Antibody
Catalog # 77701-310
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HLA-DQB1/HLA-DQB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P01920/P05538
- Antigen Synonyms:HLA-DQB2|MHC class II antigen DQB1|HLA class II histocompatibility antigen, DQ beta 1 chain|HLA class II histocompatibility antigen, DQ beta 2 chain|HLA-DQB1|HLA-DQB|HLA-DXB
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human HLA-DQB1/2
- Purification:Antigen affinity purified
- Cat. No.:77701-310
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: HLA-DQB1/HLA-DQB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human