Anti-CMTM5 Rabbit Polyclonal Antibody
Catalog # 77701-303
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CMTM5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q96DZ9
- Antigen Synonyms:CMTM5|Chemokine-like factor superfamily member 5|1500005P16Rik|2900052H21Rik|Chemokine like factor superfamily 5|Chemokine like factor super family 5|CKLFSF5|Chemokine like factor super family five variant 4|CKLF-like MARVEL transmembrane domain containing 5|FLJ37521|CKLF5_HUMAN|CKLF-like MARVEL transmembrane domain-containing protein 5
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human CMTM5
- Purification:Antigen affinity purified
- Cat. No.:77701-303
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: CMTM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human