Anti-WDR83 Rabbit Polyclonal Antibody
Catalog # 77701-264
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:WDR83
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9BRX9
- Antigen Synonyms:Mitogen activated protein kinase organizer 1
|MAPK organizer 1|WD repeat domain-containing protein 83|wdr83|MORG 1|Mitogen-activated protein kinase organizer 1|MGC4238|WDR83_HUMAN - Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human MAPK Organizer 1
- Purification:Antigen affinity purified
- Cat. No.:77701-264
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: WDR83
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat