Anti-ENPP2 Rabbit Polyclonal Antibody
Catalog # 77701-186
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ENPP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q13822
- Antigen Synonyms:ATX X|Enpp2|Phosphodiesterase I alpha|FLJ26803|ATX|Extracellular lysophospholipase D|Autotaxin t|PD IALPHA|Ectonucleotide pyrophosphatase/phosphodiesterase 2|LysoPLD|ENPP2_HUMAN|E-NPP 2|E NPP 2|Phosphodiesterase I/nucleotide pyrophosphatase 2|NPP2|Ectonucleotide pyrophosphatase/phosphodiesterase family member 2|Plasma lysophospholipase D|PDNP2|Autotaxin
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Autotaxin
- Purification:Antigen affinity purified
- Cat. No.:77701-186
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: ENPP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat