Anti-NRG1 Rabbit Polyclonal Antibody
Catalog # 77701-154
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NRG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q02297
- Antigen Synonyms:HRG1|Heregulin|NDF|nrg1|Neu differentiation factor|Pro-NRG1|heregulin, alpha|SMDF|NRG1_HUMAN|MSTP131|ARIA|NRG1-IT2|Acetylcholine receptor-inducing activity|Breast cancer cell differentiation factor p45|HRG|Glial growth factor|heregulin, alpha (45kD, ERBB2 p185-activator)|GGF2|glial growth factor 2|Acetylcholine receptor-inducing activity, chick, homolog of|MST131|GGF|Neuregulin-1|Sensory and motor neuron-derived factor|HRGA|HGL
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human Neuregulin-1
- Purification:Antigen affinity purified
- Cat. No.:77701-154
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: NRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat