Anti-GDF5 Rabbit Polyclonal Antibody
77701-138
New Product
- Antigen Symbol:GDF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P43026
- Antigen Synonyms:OS5|GDF-5|SYNS2|Cartilage-derived morphogenetic protein 1|Growth/differentiation factor 5|LAP4|CDMP1|Cartilage derived morphogenetic protein 1|GDF5_HUMAN|BMP14|Gdf5|Radotermin|CDMP-1|Growth differentiation factor 5
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human GDF-5
- Purification:Antigen affinity purified
- Cat. No.:77701-138
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: GDF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat