Anti-HOXD3 Rabbit Polyclonal Antibody
77701-035
New Product
- Antigen Symbol:HOXD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P31249
- Antigen Synonyms:Homeobox protein HoxD3|HOX4A|HOX 1D|Hox4.1|Hox 4.1|HOXD3|Hox 4A|Homeo box D3|HOX1D|MGC10470|Hox-4.1 mouse homolog of|Homeobox protein Hox-D3|HXD3_HUMAN|Homeobox protein Hox D3|HOX4|HOXD 3|Homeobox protein Hox-4A|HOX 4|HOX D3|Homeodomain protein|Homeobox D3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human HoxD3
- Purification:Antigen affinity purified
- Cat. No.:77701-035
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: HOXD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat