Anti-SFTPA2 Rabbit Polyclonal Antibody
77701-031
New Product
- Antigen Symbol:SFTPA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q8IWL1
- Antigen Synonyms:SP-A2|SFTPA2|SP-A|SFTP1|PSAP|Collectin-5|PSP-A|Alveolar proteinosis protein|SFTPA2B|SFTPA|COLEC5|Pulmonary surfactant-associated protein A2|PSPA|35 kDa pulmonary surfactant-associated protein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human SP-A
- Purification:Antigen affinity purified
- Cat. No.:77701-031
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: SFTPA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat