Anti-FGF5 Rabbit Polyclonal Antibody
Catalog # 77701-030
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FGF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P12034
- Antigen Synonyms:Fibroblast growth factor 5|heparin binding growth factor 5|FGF 5|FGF-5|Heparin-binding growth factor 5|HBGF 5|Smag 82|TCMGLY|FGF5|HBGF-5|Smag-82|FGF5_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human FGF-5
- Purification:Antigen affinity purified
- Cat. No.:77701-030
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: FGF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human