Anti-CPE Rabbit Polyclonal Antibody
77701-007
New Product
- Antigen Symbol:CPE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P16870
- Antigen Synonyms:Enkephalin convertase|CPE|Cph 1|Carboxypeptidase E|CarboxypeptidaseH|Entephalin convertase|Prohormone processing carboxypeptidase|CBPE_HUMAN|CarboxypeptidaseE|Cobalt stimulated chromaffin granule carboxypeptidase|Carboxypeptidase H precursor|Prohormone-processing carboxypeptidase|Insulin granule associated carboxypeptidase|Carboxypeptidase H|CPH
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human CPE
- Purification:Antigen affinity purified
- Cat. No.:77701-007
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: CPE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat