Anti-CCT6A Rabbit Polyclonal Antibody
Catalog # 77687-340
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CCT6A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:P40227
- Antigen Synonyms:CCT-zeta-1|CCT6A|Acute morphine dependence-related protein 2|TCP-1-zeta|Tcp20|CCT6|HTR3|T-complex protein 1 subunit zeta|CCTZ
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human T-complex protein 1 subunit zeta protein (80-250AA)
- Purification:Antigen affinity purified
- Cat. No.:77687-340
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20-1:200, IP:1:200-1:2000
Type:
Antigen: CCT6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human