Anti-CX3CR1 Rabbit Polyclonal Antibody
Catalog # 77700-954
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CX3CR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P49238
- Antigen Synonyms:C-X3-C CKR-1|Beta chemokine receptor-like 1|V28|CX3C1_HUMAN|CCRL1|CX3C CKR1|CMKDR1|CX3C chemokine receptor 1|Fractalkine receptor|CMKBLR1|CMK-BRL1|CMK BRL 1|GPR13|Chemokine C X3 C motif receptor 1|CX3CR1|C X3 C CKR 1|G-protein coupled receptor 13|CMK-BRL-1|GPRV28
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Fractalkine Receptor
- Purification:Antigen affinity purified
- Cat. No.:77700-954
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat