Anti-SLC6A14 Rabbit Polyclonal Antibody
77700-944
New Product
- Antigen Symbol:SLC6A14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9UN76
- Antigen Synonyms:Amino acid transporter ATB0+|Sodium- and chloride-dependent neutral and basic amino acid transporter B(0+|SLC6A14|Solute carrier family 6 member 14
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human SLC6A14
- Purification:Antigen affinity purified
- Cat. No.:77700-944
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: SLC6A14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human