Anti-RASSF8 Rabbit Polyclonal Antibody
Catalog # 77700-942
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RASSF8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q8NHQ8
- Antigen Synonyms:HoJ1|RASF8_HUMAN|Ras association domain containing protein 8|Ras association domain-containing protein 8|C12orf2|Ras association (RalGDS/AF 6) domain family 8|RASSF 8|Ras association domain family 8|HoJ 1|Carcinoma associated protein HOJ 1|Carcinoma-associated protein HOJ-1|Carcinoma associated HOJ 1|Rassf8|Ras association (RalGDS/AF 6) domain family (N terminal) member 8
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human RASSF8
- Purification:Antigen affinity purified
- Cat. No.:77700-942
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: RASSF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat