Anti-HGFAC Rabbit Polyclonal Antibody
77700-912
New Product
- Antigen Symbol:HGFAC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q04756
- Antigen Synonyms:EC 3.4.21.-) [Cleaved into: Hepatocyte growth factor activator short chain|Hepatocyte growth factor activator long chain]|Hepatocyte growth factor activator|HGF activator|HGFAC|HGFA
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human HGFA
- Purification:Antigen affinity purified
- Cat. No.:77700-912
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: HGFAC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat