Anti-ATP6V0D1 Rabbit Polyclonal Antibody
Catalog # 77700-869
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP6V0D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P61421
- Antigen Synonyms:V ATPase AC39 subunit|ATPase H+ transporting lysosomal (vacuolar proton pump) member D|V-ATPase AC39 subunit|ATP6V0D1|ATPase H+ transporting lysosomal V0 subunit d1|V-ATPase subunit d 1|V ATPase subunit d 1|VATX|VMA6|VA0D1_HUMAN|ATPase H+ transporting lysosomal 38kDa V0 subunit d1|Vacuolar proton pump subunit d 1|VPATPD|ATP6DV|p39|Vacuolar ATP synthase subunit d 1|ATP6D|V ATPase subunit D|VMA 6|32 kDa accessory protein|V-ATPase 40 kDa accessory protein|H(+) transporting two sector ATPase subunit D|ATPase H+ transporting lysosomal 38kD V0 subunit d|V ATPase 40 KDa accessory protein|V-type proton ATPase subunit d 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human V-ATPase D1
- Purification:Antigen affinity purified
- Cat. No.:77700-869
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ATP6V0D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat