Anti-ZC3H11A Rabbit Polyclonal Antibody
Catalog # 77700-840
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ZC3H11A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O75152
- Antigen Synonyms:Zinc finger CCCH domain-containing protein 11A|ZC3HDC11A|KIAA0663|ZC3H11A
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ZC3H11A
- Purification:Antigen affinity purified
- Cat. No.:77700-840
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: ZC3H11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey