Anti-XPA Rabbit Polyclonal Antibody
Catalog # 77700-825
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:XPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P23025
- Antigen Synonyms:xpa|XP1|Xeroderma pigmentosum group A-complementing protein|Xeroderma pigmentosum group A complementing protein|Xeroderma pigmentosum 1|DNA repair protein complementing XP-A cells|Xpac|DNA repair protein complementing XP A cells|Xeroderma pigmentosum complementation group A|DNA repair protein complementing XPA cells|Excision repair controlling|XPA_HUMAN|XP 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human XPA
- Purification:Antigen affinity purified
- Cat. No.:77700-825
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: XPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse