Anti-KLF11 Rabbit Polyclonal Antibody
Catalog # 77700-722
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KLF11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:O14901
- Antigen Synonyms:MODY7|Tieg2b|D12Ertd427e|Transforming growth factor-beta-inducible early growth response protein 3|Tcfcp2l2 |TIEG-2|Transforming Growth Factor Beta Inducible Early Growth Response 2|TIEG-3|Klf11|9830142A17|TGFB-inducible early growth response protein 2b|FKLF|TGFB-inducible early growth response protein 3|FKLF1|Krueppel-like factor 11|Krueppel like factor 11|KLF11_HUMAN|Transforming growth factor-beta-inducible early growth response protein 2|TGFB-inducible early growth response protein 2|TIEG 2|Krueppel-like transcription factor 1|TGFB Early Growth Response 2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human TIEG2
- Purification:Antigen affinity purified
- Cat. No.:77700-722
- Packaging:Vial
Specifications
About this item
Type:
Antigen: KLF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse