Anti-SOCS1 Rabbit Polyclonal Antibody
Catalog # 77700-598
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SOCS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O15524
- Antigen Synonyms:Cytokine inducible SH2 protein 1|Janus kinase binding protein|SOCS1_HUMAN|CISH1|Suppressor of cytokine signaling 1|JAK-binding protein|SOCS-1|SSI1|JAK binding protein|STAT-induced STAT inhibitor 1|SSI 1|STAT induced STAT inhibitor 1|TEC interacting protein 3|Supressor of cytokine signalling 1|TIP3|JAB|TIP 3|CISH 1|Socs1|TIP-3|SOCS 1|SSI-1|Tec-interacting protein 3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human SOCS-1
- Purification:Antigen affinity purified
- Cat. No.:77700-598
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:200-1:1000, ELISA:1:40000
Type:
Antigen: SOCS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat