Order Entry
United States
ContactUsLinkComponent
 
Anti-HSPG2 Rabbit Polyclonal Antibody
New Product
 
undefined
Anti-HSPG2 Rabbit Polyclonal Antibody
New Product

 

  • Catalog No:
  • 77708-709
  • 77708-708
  • Antigen Symbol:
  • HSPG2
  • HSPG2
  • Antigen Synonyms:
  • LG3 peptide|Basement membrane specific heparan sulfate proteoglycan core protein|SJS|Schwartz Jampel syndrome 1 (chondrodystrophic myotonia)|HSPG 2|PGBM_HUMAN|HSPG|PLC|Perlecan|Heparan sulfate proteoglycan of basement membrane|SJA|Endorepellin (domain V region)|Hspg2|SJS1
  • LG3 peptide|Basement membrane specific heparan sulfate proteoglycan core protein|SJS|Schwartz Jampel syndrome 1 (chondrodystrophic myotonia)|HSPG 2|PGBM_HUMAN|HSPG|PLC|Perlecan|Heparan sulfate proteoglycan of basement membrane|SJA|Endorepellin (domain V region)|Hspg2|SJS1
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P98160
  • P98160
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide of Human HSPG2
  • Synthetic peptide of Human HSPG2
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice.
  • Shipped on blue ice.

 

Frequently Bought Together