Anti-AMZ2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: AMZ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77708-681
- 77708-680
- Antigen Symbol:
- AMZ2
- AMZ2
- Antigen Synonyms:
- Archaemetzincin-2|Archeobacterial metalloproteinase like protein 2|Archeobacterial metalloproteinase-like protein 2|AMZ2_HUMAN|AMZ2|Archaemetzincin 2|Archaelysin family metallopeptidase 2
- Archaemetzincin-2|Archeobacterial metalloproteinase like protein 2|Archeobacterial metalloproteinase-like protein 2|AMZ2_HUMAN|AMZ2|Archaemetzincin 2|Archaelysin family metallopeptidase 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86W34
- Q86W34
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human AMZ2
- Fusion protein of Human AMZ2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together