Anti-SERPINA1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77708-656
- 77708-655
- Antigen Symbol:
- SERPINA1
- SERPINA1
- Antigen Synonyms:
- A1AT_HUMAN|PRO2275|Serpina1|AAT|Alpha 1 protease inhibitor|PRO0684|PI|Serpin A1|SPAAT|alpha1 proteinase inhibitor|Serpin peptidase inhibitor clade A member 1|Dom1|A1A|Short peptide from AAT|Serine (or cysteine) proteinase inhibitor clade A member 1|Spi1-1|Serpin A1a|Alpha 1 antitrypsin|Alpha 1 antitrypsin null|Alpha-1 protease inhibitor|PI1|A1AT|Alpha1AT|Serine protease inhibitor A1a|Alpha-1-antiproteinase|Alpha 1 antiproteinase|Serine protease inhibitor 1-1
- A1AT_HUMAN|PRO2275|Serpina1|AAT|Alpha 1 protease inhibitor|PRO0684|PI|Serpin A1|SPAAT|alpha1 proteinase inhibitor|Serpin peptidase inhibitor clade A member 1|Dom1|A1A|Short peptide from AAT|Serine (or cysteine) proteinase inhibitor clade A member 1|Spi1-1|Serpin A1a|Alpha 1 antitrypsin|Alpha 1 antitrypsin null|Alpha-1 protease inhibitor|PI1|A1AT|Alpha1AT|Serine protease inhibitor A1a|Alpha-1-antiproteinase|Alpha 1 antiproteinase|Serine protease inhibitor 1-1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01009
- P01009
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Alpha-1-antitrypsin protein (25-418AA)
- Recombinant Human Alpha-1-antitrypsin protein (25-418AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together