Order Entry
United States
ContactUsLinkComponent
Anti-POMT1 Rabbit Polyclonal Antibody
Anti-POMT1 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-POMT1 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77708-654
  • 77708-653
  • Antigen Symbol:
  • POMT1
  • POMT1
  • Antigen Synonyms:
  • EC 2.4.1.109|Dolichyl-phosphate-mannose--protein mannosyltransferase 1|Protein O-mannosyl-transferase 1|POMT1
  • EC 2.4.1.109|Dolichyl-phosphate-mannose--protein mannosyltransferase 1|Protein O-mannosyl-transferase 1|POMT1
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9Y6A1
  • Q9Y6A1
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide of Human POMT1
  • Synthetic peptide of Human POMT1
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice.
  • Shipped on blue ice.

Specifications

Frequently Bought Together