Anti-AP2S1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: AP2S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77708-561
- 77708-562
- Antigen Symbol:
- AP2S1
- AP2S1
- Antigen Synonyms:
- AP17 delta|Adaptor protein complex AP2 subunit sigma|AP2S1_HUMAN|Adaptor related protein complex 2 sigma 1 subunit|AP 2 complex subunit sigma 1|Clathrin associated/assembly/adaptor protein small 2|Adaptor protein complex AP 2 subunit sigma|Clathrin coat associated protein AP17|Clathrin coat-associated protein AP17|AI043088|Plasma membrane adaptor AP-2 17 kDa protein|Clathrin coat assembly protein AP17|Clathrin assembly protein 2 small chain|AP17|Plasma membrane adaptor AP 2 17 kDa protein|AP 17|AP-2 complex subunit sigma|CLAPS2|Clathrin adaptor protein AP17|Ap2s1|AP2 complex subunit sigma 1|sigma subunit|Plasma membrane adaptor AP2 17 kDa protein|Sigma2 adaptin|Sigma2-adaptin|Clathrin associated/assembly/adaptor protein|Sigma adaptin 3b|Adapter-related protein complex 2 sigma subunit|Adaptor protein complex AP-2 subunit sigma|small 2 (17 kD)|17-KD|Clathrin adaptor complex AP2|CLAPS 2|HA2 17 kDa subunit|MGC62945
- AP17 delta|Adaptor protein complex AP2 subunit sigma|AP2S1_HUMAN|Adaptor related protein complex 2 sigma 1 subunit|AP 2 complex subunit sigma 1|Clathrin associated/assembly/adaptor protein small 2|Adaptor protein complex AP 2 subunit sigma|Clathrin coat associated protein AP17|Clathrin coat-associated protein AP17|AI043088|Plasma membrane adaptor AP-2 17 kDa protein|Clathrin coat assembly protein AP17|Clathrin assembly protein 2 small chain|AP17|Plasma membrane adaptor AP 2 17 kDa protein|AP 17|AP-2 complex subunit sigma|CLAPS2|Clathrin adaptor protein AP17|Ap2s1|AP2 complex subunit sigma 1|sigma subunit|Plasma membrane adaptor AP2 17 kDa protein|Sigma2 adaptin|Sigma2-adaptin|Clathrin associated/assembly/adaptor protein|Sigma adaptin 3b|Adapter-related protein complex 2 sigma subunit|Adaptor protein complex AP-2 subunit sigma|small 2 (17 kD)|17-KD|Clathrin adaptor complex AP2|CLAPS 2|HA2 17 kDa subunit|MGC62945
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53680
- P53680
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human AP2S1
- Fusion protein of Human AP2S1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together