Anti-PRSS3 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PRSS3
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77708-522
- 77708-521
- Antigen Symbol:
- PRSS3
- PRSS3
- Antigen Synonyms:
- Protease, serine, 3|MTG|T9|Trypsinogen 4|Trypsin 3|Serine protease 3|PRSS3|Brain trypsinogen|Trypsin III|Protease, serine, 4 (trypsin 4, brain)|Trypsinogen 5|TRY3|Trypsin IV|Mesotrypsinogen|Trypsinogen IV|Pancreatic trypsinogen III|TRY3_HUMAN|Serine protease 4|Trypsin-3|Mesotrypsin|PRSS4|TRY4
- Protease, serine, 3|MTG|T9|Trypsinogen 4|Trypsin 3|Serine protease 3|PRSS3|Brain trypsinogen|Trypsin III|Protease, serine, 4 (trypsin 4, brain)|Trypsinogen 5|TRY3|Trypsin IV|Mesotrypsinogen|Trypsinogen IV|Pancreatic trypsinogen III|TRY3_HUMAN|Serine protease 4|Trypsin-3|Mesotrypsin|PRSS4|TRY4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35030
- P35030
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Trypsin-3 protein (81-303AA)
- Recombinant Human Trypsin-3 protein (81-303AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together