Anti-APOC3 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: APOC3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77708-434
- 77708-435
- Antigen Symbol:
- APOC3
- APOC3
- Antigen Synonyms:
- MGC150353|Apolipoprotein C3|Apolipoprotein C III|ApoC-III|APO C3|Apo-CIII|ApoCIII|APOC3_HUMAN|APOC 3|Apo CIII|Apolipoprotein C-III|APOC3|ApolipoproteinCIII|ApoC III
- MGC150353|Apolipoprotein C3|Apolipoprotein C III|ApoC-III|APO C3|Apo-CIII|ApoCIII|APOC3_HUMAN|APOC 3|Apo CIII|Apolipoprotein C-III|APOC3|ApolipoproteinCIII|ApoC III
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02656
- P02656
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Apolipoprotein C-III protein (21-99AA)
- Recombinant Human Apolipoprotein C-III protein (21-99AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together