Anti-HBA1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HBA1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77708-418
- 77708-417
- Antigen Symbol:
- HBA1
- HBA1
- Antigen Synonyms:
- Alpha 1 globin|HBA1|MGC126895|Alpha globin|MGC126897|Hemoglobin subunit alpha|Hemoglobin alpha 1|Hemoglobin alpha 2|Alpha-globin|Alpha one globin|HBA2|Hemoglobin alpha 1 globin chain|Hemoglobin alpha chain|Hemoglobin alpha 1 chain|HBA_HUMAN
- Alpha 1 globin|HBA1|MGC126895|Alpha globin|MGC126897|Hemoglobin subunit alpha|Hemoglobin alpha 1|Hemoglobin alpha 2|Alpha-globin|Alpha one globin|HBA2|Hemoglobin alpha 1 globin chain|Hemoglobin alpha chain|Hemoglobin alpha 1 chain|HBA_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P69905
- P69905
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Hemoglobin subunit alpha protein (2-142AA)
- Recombinant Human Hemoglobin subunit alpha protein (2-142AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together