Anti-SPA17 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPA17
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77708-381
- 77708-380
- Antigen Symbol:
- SPA17
- SPA17
- Antigen Synonyms:
- Sp17-1|Sperm surface protein Sp17|SPA17|SP17 1|Sperm protein 17|Cancer/testis antigen 22|CT22|Band 34|SP17_HUMAN|Sperm autoantigenic protein 17
- Sp17-1|Sperm surface protein Sp17|SPA17|SP17 1|Sperm protein 17|Cancer/testis antigen 22|CT22|Band 34|SP17_HUMAN|Sperm autoantigenic protein 17
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15506
- Q15506
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Sperm surface protein Sp17 protein (1-146AA)
- Recombinant Human Sperm surface protein Sp17 protein (1-146AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together