Anti-TRIB3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77708-320
- 77708-321
- Antigen Symbol:
- TRIB3
- TRIB3
- Antigen Synonyms:
- p65 interacting inhibitor of NF-kappaB|Neuronal cell death inducible putative kinase|Trib3|TRIB 3|Tribbles3|SKIP 3|TRB 3|SINK|SKIP3|TRIB3_HUMAN|C20orf97|Tribbles homolog 3|TRB3|Neuronal cell death-inducible putative kinase|p65-interacting inhibitor of NF-kappa-B|NIPK|Tribbles pseudokinase 3|TRB-3
- p65 interacting inhibitor of NF-kappaB|Neuronal cell death inducible putative kinase|Trib3|TRIB 3|Tribbles3|SKIP 3|TRB 3|SINK|SKIP3|TRIB3_HUMAN|C20orf97|Tribbles homolog 3|TRB3|Neuronal cell death-inducible putative kinase|p65-interacting inhibitor of NF-kappa-B|NIPK|Tribbles pseudokinase 3|TRB-3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96RU7
- Q96RU7
- Isotype:
- IgG
- IgG
- Immunogen:
- Full length fusion protein
- Full length fusion protein
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together