Anti-CCL25 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: CCL25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77708-217
- 77708-216
- Antigen Symbol:
- CCL25
- CCL25
- Antigen Synonyms:
- SCYA 25|CCL25|CCL25_HUMAN|Ck beta 15|MGC125074|Small inducible cytokine subfamily A Cys Cys member 25|member 25|Small inducible cytokine subfamily A|Ckb 15|Thymus-expressed chemokine|Small inducible cytokine A25|CCL 25|Chemokine CC motif ligand 25|C-C motif chemokine 25|Chemokine (C C motif) ligand 25|Small-inducible cytokine A25|Small inducible cytokine A25 isoform 1 precursor|Small inducible cytokine A25 isoform 2 precursor|TECK|Small inducible cytokine subfamily A (Cys Cys) member 25|Chemokine (CC motif) ligand 25|TECKvar|SCY A25|SCYA25|Thymus expressed chemokine|Ckb15|MGC150327|Chemokine C C motif ligand 25|Chemokine TECK|A130072A22Rik
- SCYA 25|CCL25|CCL25_HUMAN|Ck beta 15|MGC125074|Small inducible cytokine subfamily A Cys Cys member 25|member 25|Small inducible cytokine subfamily A|Ckb 15|Thymus-expressed chemokine|Small inducible cytokine A25|CCL 25|Chemokine CC motif ligand 25|C-C motif chemokine 25|Chemokine (C C motif) ligand 25|Small-inducible cytokine A25|Small inducible cytokine A25 isoform 1 precursor|Small inducible cytokine A25 isoform 2 precursor|TECK|Small inducible cytokine subfamily A (Cys Cys) member 25|Chemokine (CC motif) ligand 25|TECKvar|SCY A25|SCYA25|Thymus expressed chemokine|Ckb15|MGC150327|Chemokine C C motif ligand 25|Chemokine TECK|A130072A22Rik
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15444
- O15444
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CCL25
- Fusion protein of Human CCL25
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Frequently Bought Together