Anti-PLAT Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77708-172
- 77708-171
- Antigen Symbol:
- PLAT
- PLAT
- Antigen Synonyms:
- t-PA|Plasminogen activator tissue type|Alteplase|DKFZp686I03148|T Plasminogen Activator|TPA1|t PA|Reteplase|Tissue plasminogen activator (t PA)|PLAT|Plasminogen activator tissue|T-plasminogen activator|Tissue type plasminogen activator|Tissue-type plasminogen activator chain B|tPA|TPA_HUMAN
- t-PA|Plasminogen activator tissue type|Alteplase|DKFZp686I03148|T Plasminogen Activator|TPA1|t PA|Reteplase|Tissue plasminogen activator (t PA)|PLAT|Plasminogen activator tissue|T-plasminogen activator|Tissue type plasminogen activator|Tissue-type plasminogen activator chain B|tPA|TPA_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P00750
- P00750
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Tissue-type plasminogen activator protein (36-556AA)
- Recombinant Human Tissue-type plasminogen activator protein (36-556AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together