Anti-PDCD2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: PDCD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77708-069
- 77708-070
- Antigen Symbol:
- PDCD2
- PDCD2
- Antigen Synonyms:
- Zinc finger protein Rp 8|Zinc finger MYND domain-containing protein 7|OTTHUMP00000017702|PDCD2_HUMAN|Zinc finger MYND domain containing protein 7|ZMYND7|Programmed cell death 2|Zinc finger protein Rp-8|PDCD2 protein|PDCD 2|Rp8 homolog|RP8|Programmed cell death protein 2|Pdcd2|MGC12347|Zinc finger protein Rp8|ZMYND 7|RP 8
- Zinc finger protein Rp 8|Zinc finger MYND domain-containing protein 7|OTTHUMP00000017702|PDCD2_HUMAN|Zinc finger MYND domain containing protein 7|ZMYND7|Programmed cell death 2|Zinc finger protein Rp-8|PDCD2 protein|PDCD 2|Rp8 homolog|RP8|Programmed cell death protein 2|Pdcd2|MGC12347|Zinc finger protein Rp8|ZMYND 7|RP 8
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16342
- Q16342
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human PDCD2
- Synthetic peptide of Human PDCD2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together