Anti-PAK1 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:5000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: PAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77708-004
- 77708-005
- Antigen Symbol:
- PAK1
- PAK1
- Antigen Synonyms:
- ADRB2|Protein kinase MUK2|PAKalpha|p21/Cdc42/Rac1 activated kinase 1 (yeast Ste20 related)|Rac/p21-activated kinase|p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast)|p65-PAK|Alpha PAK|MGC130000|PAK alpha|STE20 homolog yeast|p65 PAK|Alpha-PAK|p21 protein (Cdc42/Rac) activated kinase 1|p21-activated kinase 1|Paka|MGC130001|p21 activated kinase 1|Pak1|PAK-1|PAK1_HUMAN|P68-PAK|Serine/threonine-protein kinase PAK 1
- ADRB2|Protein kinase MUK2|PAKalpha|p21/Cdc42/Rac1 activated kinase 1 (yeast Ste20 related)|Rac/p21-activated kinase|p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast)|p65-PAK|Alpha PAK|MGC130000|PAK alpha|STE20 homolog yeast|p65 PAK|Alpha-PAK|p21 protein (Cdc42/Rac) activated kinase 1|p21-activated kinase 1|Paka|MGC130001|p21 activated kinase 1|Pak1|PAK-1|PAK1_HUMAN|P68-PAK|Serine/threonine-protein kinase PAK 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13153
- Q13153
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human PAK1
- Synthetic peptide of Human PAK1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Frequently Bought Together