Anti-CLEC2D Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: CLEC2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77707-885
- 77707-884
- Antigen Symbol:
- CLEC2D
- CLEC2D
- Antigen Synonyms:
- CLC2D_HUMAN|LLT1|Lectin-like NK cell receptor|CLEC2D protein|Osteoclast inhibitory lectin|member D|CLAX|Lectin like NK cell receptor|CLEC2D|OCIL|Lectin like transcript 1|C type lectin superfamily 2|Lectin-like transcript 1|LLT-1|Clec2d5|L LT1|C-type lectin domain family 2 member D|C type lectin related f
- CLC2D_HUMAN|LLT1|Lectin-like NK cell receptor|CLEC2D protein|Osteoclast inhibitory lectin|member D|CLAX|Lectin like NK cell receptor|CLEC2D|OCIL|Lectin like transcript 1|C type lectin superfamily 2|Lectin-like transcript 1|LLT-1|Clec2d5|L LT1|C-type lectin domain family 2 member D|C type lectin related f
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHP7
- Q9UHP7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CLEC2D
- Fusion protein of Human CLEC2D
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Frequently Bought Together