Anti-IFNAR2 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IFNAR2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77707-836
- 77707-835
- Antigen Symbol:
- IFNAR2
- IFNAR2
- Antigen Synonyms:
- IFNARB|IFN-alpha/beta receptor 2|Type I interferon receptor 2|IFN-a/脽R脽|Interferon alpha/beta receptor 2|IFN-alpha binding protein|IFN-a/脽R2|IFN alpha REC|IFNR|IFNABR|IFNAR2|Interferon alpha/beta receptor beta chain|Interferon (alpha beta and omega) receptor 2|Type I interferon receptor|INAR2_HUMAN|Interferon alpha binding protein|IFN R|IFN-R-2|Human interferon alpha/beta receptor
- IFNARB|IFN-alpha/beta receptor 2|Type I interferon receptor 2|IFN-a/脽R脽|Interferon alpha/beta receptor 2|IFN-alpha binding protein|IFN-a/脽R2|IFN alpha REC|IFNR|IFNABR|IFNAR2|Interferon alpha/beta receptor beta chain|Interferon (alpha beta and omega) receptor 2|Type I interferon receptor|INAR2_HUMAN|Interferon alpha binding protein|IFN R|IFN-R-2|Human interferon alpha/beta receptor
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48551
- P48551
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon alpha/beta receptor 2 protein (27-243AA)
- Recombinant Human Interferon alpha/beta receptor 2 protein (27-243AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together