Anti-SEPT4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: SEPT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77707-736
- 77707-735
- Antigen Symbol:
- SEPT4
- SEPT4
- Antigen Synonyms:
- CE5B3|hCDCREL-2|SEP 4|Bradeion|Septin M|MART|Apoptosis-related protein in the TGF-beta signaling pathway|SEPT4_HUMAN|Cerebral protein 7|hucep-7|CE5B3 beta|Septin-4|SEP4|H5|Peanut-like protein 2|Brain protein H5|Cell division control-related protein 2|ARTS|Bradeion beta|PNUTL2|Sept4
- CE5B3|hCDCREL-2|SEP 4|Bradeion|Septin M|MART|Apoptosis-related protein in the TGF-beta signaling pathway|SEPT4_HUMAN|Cerebral protein 7|hucep-7|CE5B3 beta|Septin-4|SEP4|H5|Peanut-like protein 2|Brain protein H5|Cell division control-related protein 2|ARTS|Bradeion beta|PNUTL2|Sept4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43236
- O43236
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SEPT4
- Synthetic peptide of Human SEPT4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together