Order Entry
United States
ContactUsLinkComponent
Anti-MUC15 Rabbit Polyclonal Antibody
Anti-MUC15 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MUC15 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77707-725
  • 77707-724
  • Antigen Symbol:
  • MUC15
  • MUC15
  • Antigen Synonyms:
  • Mucin 15|MUC 15|PAS3|Glycoprotein C|Mucin 15 cell surface associated|Mucin-15|MUC-15|MUC15_HUMAN|Component II|Muc15|Glycoprotein 4|PASIII
  • Mucin 15|MUC 15|PAS3|Glycoprotein C|Mucin 15 cell surface associated|Mucin-15|MUC-15|MUC15_HUMAN|Component II|Muc15|Glycoprotein 4|PASIII
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q8N387
  • Q8N387
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Fusion protein of Human MUC15
  • Fusion protein of Human MUC15
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice.
  • Shipped on blue ice.

Specifications

Frequently Bought Together