Anti-ACPP Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: ACPP
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77707-718
- 77707-719
- Antigen Symbol:
- ACPP
- ACPP
- Antigen Synonyms:
- Ecto-5'-nucleotidase|Ecto 5' nucleotidase|5' nucleotidase|5'-NT|ACP3|5' NT|Acid phosphatase prostate|Thiamine monophosphatase|PPAP_HUMAN|PAP|Prostatic acid phosphotase|Prostatic acid phosphatase|5'-nucleotidase|acpP|ACP 3|TMPase
- Ecto-5'-nucleotidase|Ecto 5' nucleotidase|5' nucleotidase|5'-NT|ACP3|5' NT|Acid phosphatase prostate|Thiamine monophosphatase|PPAP_HUMAN|PAP|Prostatic acid phosphotase|Prostatic acid phosphatase|5'-nucleotidase|acpP|ACP 3|TMPase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15309
- P15309
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Prostatic acid phosphatase protein (38-386AA)
- Recombinant Human Prostatic acid phosphatase protein (38-386AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Frequently Bought Together