Anti-ABCB7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: ABCB7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77707-461
- 77707-460
- Antigen Symbol:
- ABCB7
- ABCB7
- Antigen Synonyms:
- Atm1p|ATP-binding cassette transporter 7|ATP binding cassette sub family B member 7 mitochondrial|ATP binding cassette 7|ABCB7_HUMAN|Abcb7|mitochondrial|ABC transporter 7 protein|ATP binding cassette transporter 7|PGP7|ATP-binding cassette sub-family B member 7|ATP binding cassette sub family B member 7|ABC7|ASAT|MDR7|Multidrug resistance protein 7|P-glycoprotein 7|EST140535|ATP binding cassette sub family B (MDR/TAP) member 7
- Atm1p|ATP-binding cassette transporter 7|ATP binding cassette sub family B member 7 mitochondrial|ATP binding cassette 7|ABCB7_HUMAN|Abcb7|mitochondrial|ABC transporter 7 protein|ATP binding cassette transporter 7|PGP7|ATP-binding cassette sub-family B member 7|ATP binding cassette sub family B member 7|ABC7|ASAT|MDR7|Multidrug resistance protein 7|P-glycoprotein 7|EST140535|ATP binding cassette sub family B (MDR/TAP) member 7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75027
- O75027
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ABCB7
- Fusion protein of Human ABCB7
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together