Anti-NCEH1 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:100-1:300
Type:
Antigen: NCEH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77707-440
- 77707-441
- Antigen Symbol:
- NCEH1
- NCEH1
- Antigen Synonyms:
- Arylacetamide deacetylase like 1|NCEH|Neutral cholesterol ester hydrolase|Neutral cholesterol ester hydrolase 1|Nceh1|NCEH1_HUMAN|Arylacetamide deacetylase-like 1
- Arylacetamide deacetylase like 1|NCEH|Neutral cholesterol ester hydrolase|Neutral cholesterol ester hydrolase 1|Nceh1|NCEH1_HUMAN|Arylacetamide deacetylase-like 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6PIU2
- Q6PIU2
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human NCEH1
- Fusion protein of Human NCEH1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Frequently Bought Together