Anti-DNAJB4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: DNAJB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77707-430
- 77707-429
- Antigen Symbol:
- DNAJB4
- DNAJB4
- Antigen Synonyms:
- HLJ1|HSP40 homolog|DnaJ like heat shock protein 40|HLJ 1|DNJB4_HUMAN|Heat shock protein 40 homolog|Heat shock 40 kDa protein 1 homolog|DNAJB 4|DNAJB4|DnaJ (Hsp40) homolog subfamily B member 4|DnaJ homolog subfamily B member 4|DNAJW|DjB4|Human liver DnaJ-like protein
- HLJ1|HSP40 homolog|DnaJ like heat shock protein 40|HLJ 1|DNJB4_HUMAN|Heat shock protein 40 homolog|Heat shock 40 kDa protein 1 homolog|DNAJB 4|DNAJB4|DnaJ (Hsp40) homolog subfamily B member 4|DnaJ homolog subfamily B member 4|DNAJW|DjB4|Human liver DnaJ-like protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UDY4
- Q9UDY4
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human DnaJ homolog subfamily B member 4 protein (1-337AA)
- Recombinant Human DnaJ homolog subfamily B member 4 protein (1-337AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together