Order Entry
United States
ContactUsLinkComponent
Anti-MYL2 Rabbit Polyclonal Antibody
Anti-MYL2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MYL2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77707-249
  • 77707-248
  • Antigen Symbol:
  • MYL2
  • MYL2
  • Antigen Synonyms:
  • Myosin regulatory light chain 2|MYL2|CMH10|MLC 2v|ventricular/cardiac muscle isoform|MYL 2|Slow cardiac myosin regulatory light chain 2|Myosin light polypeptide 2 regulatory cardiac slow|MLC2|RLC of myosin|Myosin regulatory light chain 2 ventricular/cardiac muscle isoform|MLRV_HUMAN|MLC-2|Regulatory light chain of myosin|MLC-2v|Cardiac myosin light chain-2|Cardiac ventricular myosin light chain 2|Myosin light chain 2 regulatory cardiac slow
  • Myosin regulatory light chain 2|MYL2|CMH10|MLC 2v|ventricular/cardiac muscle isoform|MYL 2|Slow cardiac myosin regulatory light chain 2|Myosin light polypeptide 2 regulatory cardiac slow|MLC2|RLC of myosin|Myosin regulatory light chain 2 ventricular/cardiac muscle isoform|MLRV_HUMAN|MLC-2|Regulatory light chain of myosin|MLC-2v|Cardiac myosin light chain-2|Cardiac ventricular myosin light chain 2|Myosin light chain 2 regulatory cardiac slow
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P10916
  • P10916
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Fusion protein of Human MYL2
  • Fusion protein of Human MYL2
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice.
  • Shipped on blue ice.

Specifications

Frequently Bought Together