Anti-LSM10 Rabbit Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: LSM10
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77720-466
- 77720-465
- Antigen Symbol:
- LSM10
- LSM10
- Antigen Synonyms:
- LSM10U7 snRNA-associated Sm-like protein LSm10
- LSM10U7 snRNA-associated Sm-like protein LSm10
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969L4
- Q969L4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human U7 snRNA-associated Sm-like protein LSm10 protein (1-123AA)
- Recombinant Human U7 snRNA-associated Sm-like protein LSm10 protein (1-123AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together