Anti-RND1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:200-1:500, IF:1:50-1:200
Type:
Antigen: RND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-382
- 77720-381
- Antigen Symbol:
- RND1
- RND1
- Antigen Synonyms:
- Ras homolog gene family member S|ARHS|Rho family GTPase 1|Rho-related GTP-binding protein Rho6|RND 1|RHOS|RHO 6|RHO S|Rnd1|FLJ42294|Rho related GTP binding protein Rho6|RHO6|RND1_HUMAN|GTP binding protein RHO6
- Ras homolog gene family member S|ARHS|Rho family GTPase 1|Rho-related GTP-binding protein Rho6|RND 1|RHOS|RHO 6|RHO S|Rnd1|FLJ42294|Rho related GTP binding protein Rho6|RHO6|RND1_HUMAN|GTP binding protein RHO6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92730
- Q92730
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Rho-related GTP-binding protein Rho6 protein (1-106AA)
- Recombinant Human Rho-related GTP-binding protein Rho6 protein (1-106AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together