Anti-NDRG1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDRG1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-360
- 77720-361
- Antigen Symbol:
- NDRG1
- NDRG1
- Antigen Synonyms:
- Nickel-specific induction protein Cap43|Differentiation related gene1 protein|TDD5|Reducing agents and tunicamycin responsive protein|Ndr 1|Reducing agents and tunicamycin-responsive protein|drg1|Protein regulated by oxygen1|ndr1|DRG-1|GC4|cap43|Rit42|Nmyc downstream regulated gene1|Reducin|gc4|NDRG1_HUMAN|Differentiation-related gene 1 protein|Anti GC4|hmsnl|Drg 1|Reduced in tumor|Nmyc downstream regulated|Proxy1|cmt4d|N myc downstream regulated gene 1 protein|42 kDa|Tunicamycin responsive protein|Ndrg1|nmsl|NDRG1 protein|targ1|Protein NDRG1|Protein regulated by oxygen 1|Human mRNA for RTP complete cds|Nmyc downstream regulated gene1 protein|N myc downstream regulated gene 1|RTP|N-myc downstream-regulated gene 1 protein|tdds|NDRG 1|Nickel specific induction protein|Nickel specific induction protein Cap43
- Nickel-specific induction protein Cap43|Differentiation related gene1 protein|TDD5|Reducing agents and tunicamycin responsive protein|Ndr 1|Reducing agents and tunicamycin-responsive protein|drg1|Protein regulated by oxygen1|ndr1|DRG-1|GC4|cap43|Rit42|Nmyc downstream regulated gene1|Reducin|gc4|NDRG1_HUMAN|Differentiation-related gene 1 protein|Anti GC4|hmsnl|Drg 1|Reduced in tumor|Nmyc downstream regulated|Proxy1|cmt4d|N myc downstream regulated gene 1 protein|42 kDa|Tunicamycin responsive protein|Ndrg1|nmsl|NDRG1 protein|targ1|Protein NDRG1|Protein regulated by oxygen 1|Human mRNA for RTP complete cds|Nmyc downstream regulated gene1 protein|N myc downstream regulated gene 1|RTP|N-myc downstream-regulated gene 1 protein|tdds|NDRG 1|Nickel specific induction protein|Nickel specific induction protein Cap43
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92597
- Q92597
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein NDRG1 protein (136-394AA)
- Recombinant Human Protein NDRG1 protein (136-394AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together